missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Oxide, ReagentPlus™, Honeywell™

Product Code. p-7071819 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
This item is not returnable. View return policy

Product Code. 15601390

Brand: Honeywell Research Chemicals 140211KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Quantity 1 kg
Linear Formula TiO2
UN Number NONH for all modes of transport
Formula Weight 79.87g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Density 4.26g/mL at 25°C
Chemical Name or Material Titanium(Iv) Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.