missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Oxide, ReagentPlus™, Honeywell™

Product Code. 15611390 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15611390 25 kg 25kg
15601390 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15611390 Supplier Honeywell Research Chemicals Supplier No. 1402125KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Quantity 25 kg
Linear Formula TiO2
UN Number NONH for all modes of transport
Formula Weight 79.87g/mol
Percent Purity ≥99%
Grade ReagentPlus
Density 4.26g/mL at 25°C
Chemical Name or Material Titanium(Iv) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.