missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, high surface area, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11338428
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
13463-67-7
O2Ti
79.87
MFCD00011269 MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O
This item is not returnable. View return policy
13463-67-7
O2Ti
79.87
MFCD00011269 MFCD00210650
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
26042
CHEBI:32234
dioxotitanium
O=[Ti]=O

Photocatalyst, catalyst, catalyst supportTitanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2500°C to 3000°C
Melting Point 1830°C to 1850°C
Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.9% metal basis
Solubility Information Insoluble in water,HCl,HNO3,dilute H2SO4; soluble in hot,conc. H2SO4,HF,and alkaliesSlightly soluble in hydrofluoric acid and hot concentrated sulfuric acid. Insoluble in water,hydrochloric acid,dilute sulfuric acid,and organic solvents.
Formula Weight 79.9
Odor Odorless
Density 4.26
Chemical Name or Material Titanium(IV) oxide, High surface Area: S.A. 180-300 m2/g
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'