missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, catalyst support

Product Code. 11320488
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11320488 1 kg 1kg
11300488 250 g 250g
11310488 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11320488 Supplier Thermo Scientific Alfa Aesar Supplier No. 043828.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-70-0 | O2Ti | 79.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Pellets
Quantity 1 kg
Formula Weight 79.9
Packaging Plastic bottle
Chemical Name or Material Titanium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.