missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion

Product Code. 11485250
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485250 1 kg 1kg
11475250 500 g 500g
11495250 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485250 Supplier Thermo Scientific Alfa Aesar Supplier No. 044517.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

UV protection in clear coatings | CAS: 13463-67-7 | O2Ti | 79.87 g/mol

UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 368°C
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 79.9
Concentration or Composition (by Analyte or Components) 20 to 35% in H2O
Density 1.26 g/mL
Chemical Name or Material Titanium (IV) oxide, Colloidal dispersion

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.