missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) isopropoxide, 99.995% (metals basis)

Product Code. 11368687
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11368687

Brand: Thermo Scientific Alfa Aesar 035794.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 546-68-9 | C12H28O4Ti | 284.219 g/mol

Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of Sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.219
MDL Number MFCD00008871
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Melting Point 16°C to 20°C
Color Colorless
Density 0.955 g/mL
Boiling Point 232°C
Flash Point 46°C (115°F)
Linear Formula Ti[OCH(CH3)2]4
Refractive Index 1.464
Quantity 25 g
UN Number UN2413
Beilstein 3679474
Sensitivity Moisture sensitive
Merck Index 14,9480
Solubility Information Miscible with anhydrous ethanol,chloroform,ether and benzene.
Formula Weight 284.23
Percent Purity 99.995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Titanium(IV) isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) isopropoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetraisopropyl titanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.