missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) fluoride, 99%, Thermo Scientific Chemicals

Product Code. 10658064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10658064 100 g Plastic bottle 100g
10003700 10 g Plastic bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10658064 Supplier Thermo Scientific Acros Supplier No. 308431000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-63-3
Molecular Formula TiF4
Molecular Weight (g/mol) 123.89
MDL Number MFCD00042545

Specifications

Color White
Physical Form Chunks and/or Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula TiF4
Merck Index 15, 9635
Formula Weight 123.89
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Titanium(IV) fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.