Learn More
Titanium(IV) fluoride, 99%, ACROS Organics™
Brand: Acros Organics 308430100
Code : 29
Additional Details : CAS Number : 7783-63-3 Weight : 0.06000kg Transport : UN number : 3260 Chem class : 8 Pack group : II
Packaging | Plastic bottle |
---|---|
Quantity | 10g |
Description
Lewis acidChemical Identifiers
7783-63-3 | |
TiF4 | |
123.89 | |
MFCD00042545 |
Specifications
Titanium(IV) fluoride | |
99% | |
TiF4 | |
15, 9635 | |
123.89 | |
99% | |
White |
7783-63-3 | |
TiF4 | |
MFCD00042545 | |
123.89 | |
Powder and/or Chunks | |
Plastic bottle | |
10g |