missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) fluoride, 98%, Thermo Scientific Chemicals

Catalog Number p-4904559
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7783-63-3
TiF4
MFCD00042545
This item is not returnable. View return policy
7783-63-3
TiF4
MFCD00042545

Titanium(IV) fluoride is used for preparation of glycosyl fluorides, glycosylation, preparation of fluorohydrins, addition of carbanions to aldehydes and imines. It is a catalyst for chemoselective synthesis and deprotection of geminal diacetates of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 284° (sublimation)
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98%
Solubility Information Insoluble in water.
Formula Weight 123.89
Odor Odorless
Sensitivity Moisture sensitive
Density 2.798
Chemical Name or Material Titanium(IV) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'