missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) fluoride, 98%

Product Code. 11357237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357237 100 g 100g
11347237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357237 Supplier Thermo Scientific Alfa Aesar Supplier No. 011540.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-63-3 | TiF4

Titanium(IV) fluoride is used for preparation of glycosyl fluorides, glycosylation, preparation of fluorohydrins, addition of carbanions to aldehydes and imines. It is a catalyst for chemoselective synthesis and deprotection of geminal diacetates of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-63-3
Molecular Formula TiF4
MDL Number MFCD00042545

Specifications

Melting Point 284° (sublimation)
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98%
Solubility Information Insoluble in water.
Formula Weight 123.89
Odor Odorless
Sensitivity Moisture sensitive
Density 2.798
Chemical Name or Material Titanium(IV) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.