missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide, TiO{2} 33% min

Product Code. 15412967
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 15412967

Brand: Thermo Scientific Alfa Aesar 077142.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3087-36-3 | C8H20O4Ti | 228.111 g/mol

Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.111
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Boiling Point 150°C to 152°C (10mmHg)
Melting Point -40°C
Color Yellow
Physical Form Liquid
Quantity 250 g
Linear Formula Ti(OCH2CH3)4
UN Number UN2924
Flash Point 28°C (84°F)
Solubility Information Soluble in organic solvents; Decomposes in water.
Formula Weight 228.15
Concentration or Composition (by Analyte or Components) TiO2 33% min
Odor Alcohol-like
Refractive Index 1.5043
Sensitivity Moisture sensitive
Density 1.088 g/mL
Chemical Name or Material Titanium (IV) ethoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) ethoxide, TiO2 33% min
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : Titanium(IV) ethoxide, Titanium(IV) isopropoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.