missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Chloride Solution, 1.0M in methylene chloride, Honeywell™

Product Code. p-7120877 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Unit Size:
100mL
800mL
Product Code. Quantity Packaging unitSize
15652710 100 mL Glass Bottle 100mL
15662710 800 mL Glass Bottle 800mL
2 options
This item is not returnable. View return policy

Product Code. 15652710

Brand: Honeywell-Fluka 249866100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

1.0 M in methylene chloride

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Quantity 100 mL
Linear Formula TiCl4
UN Number UN3289
Formula Weight 189.68g/mol
Concentration or Composition (by Analyte or Components) 1.0M in methylene chloride
Packaging Glass Bottle
Vapor Pressure 24.09psi (55°C), 6.93psi (20°C)
Density 1.365g/mL at 25°C
Chemical Name or Material Titanium(IV) Chloride Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.