missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride solution, 0.09 M in 20% HCl, Honeywell Fluka™

Product Code. 15655430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
4 x 100 mL
Packaging:
Glass Bottle
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655430 4 x 100 mL Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15655430 Supplier Honeywell-Fluka Supplier No. 4049854X100ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7550-45-0
Quantity 4 x 100 mL
Molecular Formula Cl4Ti
Linear Formula H2O
MDL Number MFCD00011332
UN Number UN2922
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
IUPAC Name titanium(4+);tetrachloride
Molecular Weight (g/mol) 189.667
PubChem CID 160960
Packaging Glass Bottle
Density 1.099 g/mL at 25 °C
Chemical Name or Material Titanium(IV) chloride solution, 0.09 M in 20% HCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.