missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Chloride, ≥99.995% trace metals basis, Honeywell™

Product Code. 15602750 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15602750 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15602750 Supplier Honeywell Research Chemicals Supplier No. 25431210G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99.995% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 136.4°C
Melting Point -25°C
CAS 7550-45-0
Quantity 10 g
Molecular Formula Cl4Ti
Linear Formula TiCl4
MDL Number MFCD00011267
UN Number UN1838
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
IUPAC Name titanium(4+);tetrachloride
Molecular Weight (g/mol) 189.667
PubChem CID 160960
Formula Weight 189.68g/mol
Percent Purity ≥99.995% (trace metals basis)
Packaging Glass Bottle
Vapor Pressure 50mmHg (55°C), 9.6mmHg (20°C)
Density 1.73g/mL at 20°C
Chemical Name or Material Titanium(IV) Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.