missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride, 99.99% (metals basis)

Product Code. 15441117
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441117 100 g 100g
15431117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441117 Supplier Thermo Scientific Alfa Aesar Supplier No. 022979.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol

Titanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Boiling Point 136°C to 137°C
Melting Point °C to 25°C
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Miscible with ethanol,hydrochloric acid and organic solvents.
Formula Weight 189.69
Refractive Index 1.61
Sensitivity Moisture sensitive
Density 1.726 g/mL
Chemical Name or Material Titanium(IV) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.