missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Product Code. 11374507
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374507 50 g 50g
11364507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374507 Supplier Thermo Scientific Alfa Aesar Supplier No. 039592.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Titanium(IV) bromide is used as a Lewis acid catalyst in organic synthesis. It reacts with tetrahydrofurnan and 2-methylpyridine and forms adducts such as TiBr4(THF)2 and TiBr4(2-MePy). It is also used for the formation of titanium silicide thin films on silicon substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
MDL Number MFCD00011265

Specifications

Color Orange
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98%
Formula Weight 367.54
Odor Acrid
Chemical Name or Material Titanium(IV) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.