missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Catalog Number 11364507
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7789-68-6
TiBr4
MFCD00011265
This item is not returnable. View return policy
7789-68-6
TiBr4
MFCD00011265

Titanium(IV) bromide is used as a Lewis acid catalyst in organic synthesis. It reacts with tetrahydrofurnan and 2-methylpyridine and forms adducts such as TiBr4(THF)2 and TiBr4(2-MePy). It is also used for the formation of titanium silicide thin films on silicon substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 233°C
Melting Point 39°C
Color Orange
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Solubility Information Soluble in absolute alcohol. Decomposes in waterSoluble in chlorocarbons,benzene and alcohol.
Formula Weight 367.54
Odor Acrid
Sensitivity Moisture sensitive
Density 2.6
Chemical Name or Material Titanium(IV) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'