missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) 2-ethylhexanoate, 97%

Product Code. 11435390
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435390 500 g 500g
11425390 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435390 Supplier Thermo Scientific Alfa Aesar Supplier No. 044602.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3645-34-9 | C32H60O8Ti | 620.69 g/mol

Titanium(IV) 2-ethylhexanoate acts as a coupling agent to improve the adhesion of the fiber and resin in Kevla-phenolic composites. It is used as a catalyst in the polymerization of cyclic poly(butylene terephthalate) oligomers for composite applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3645-34-9
Molecular Formula C32H60O8Ti
Molecular Weight (g/mol) 620.69
MDL Number MFCD05663781
InChI Key GDKAXSGMPFSRJY-UHFFFAOYNA-J
Synonym titanium iv 2-ethylhexanoate, hexanoic acid, 2-ethyl-, titanium 4+ salt, titanium ethylhexanoate, hexanoic acid, 2-ethyl-, titanium 4+ salt 4:1, 2-ethylhexanoate; titanium 4+, titanium iv 2-ethylhexanoate 100g, tetrakis 2-ethylhexanoic acid titanium iv salt, titanium 4+ tetrakis 2-ethylhexanoate
PubChem CID 6451578
IUPAC Name 2-ethylhexanoate;titanium(4+)
SMILES [Ti+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Specifications

Odor Odorless
Quantity 500 g
Solubility Information Immiscible with water. Miscible with ethyl acetate.
Formula Weight 623.75
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Titanium (IV) 2-ethylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.