missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid

Product Code. 10253970 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10253970 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10253970 Supplier Fisher Chemical Supplier No. T/2000/PB08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

O12S3Ti2, CAS Number-10343-61-0, 69hci1s2xi, acmc-1bolh, dititanium tris sulphate, sulfuric acid, titanium 3+ salt 3:2, titan iii-sulfat, titanium 3+ sulfate 2:3, titanium iii sulfate, titanium sesquisulfate, unii-69hci1s2xi, 500mL, Purple, SOBXOQKKUVQETK-UHFFFAOYSA-H, titanium(3+);trisulfate, 36317

TRUSTED_SUSTAINABILITY

Specifications

Color Purple
CAS 10343-61-0
Molecular Formula O12S3Ti2
MDL Number 36317
Synonym titanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
InChI Key SOBXOQKKUVQETK-UHFFFAOYSA-H
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]
IUPAC Name titanium(3+);trisulfate
Molecular Weight (g/mol) 383.902
PubChem CID 165866
Packaging Plastic Coated Glass bottle
pH 1
Quantity 500 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(III) sulfate solution, about 15% w/v
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid, Sulfuric acid, titanium(3+) salt (3:2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.