missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) sulfate, 20% in 1-4% sulfuric acid

Product Code. p-7050891
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11334617

Brand: Thermo Scientific Alfa Aesar 039664.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10343-61-0 | O12S3Ti2 | 383.902 g/mol

Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10343-61-0
Molecular Formula O12S3Ti2
Molecular Weight (g/mol) 383.902
MDL Number MFCD00036317
InChI Key SOBXOQKKUVQETK-UHFFFAOYSA-H
Synonym titanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
PubChem CID 165866
IUPAC Name titanium(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]

Specifications

Physical Form Liquid
Quantity 250 g
UN Number UN3264
Merck Index 14,9476
Solubility Information Fully miscible with water.
Formula Weight 383.38
Concentration or Composition (by Analyte or Components) 20% in 1 to 4% sulfuric acid
Density 1.15 g/mL
Chemical Name or Material Titanium(III) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.