missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(III) sulfate, 20% in 1-4% sulfuric acid, Thermo Scientific Chemicals
266.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 5 kg |
UN Number | UN3264 |
Merck Index | 14,9476 |
Solubility Information | Fully miscible with water. |
Formula Weight | 383.38 |
Concentration or Composition (by Analyte or Components) | 20% in 1 to 4% sulfuric acid |
Density | 1.15 g/mL |
Chemical Name or Material | Titanium(III) sulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'