missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) oxide, 99.8% (metals basis)

Product Code. 11364889
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364889 50 g 50g
11374889 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364889 Supplier Thermo Scientific Alfa Aesar Supplier No. 077127.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1344-54-3 | O3Ti2 | 143.731 g/mol

Titanium(III) oxide is used to study the bioacceptability of medical implant surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-54-3
Molecular Formula O3Ti2
Molecular Weight (g/mol) 143.731
MDL Number MFCD00036278
InChI Key GQUJEMVIKWQAEH-UHFFFAOYSA-N
Synonym titanium iii oxide, oxo oxotitaniooxy titanium, oxo-oxotitaniooxy titanium, titanium sesquioxide, titatnum iii oxide, ti2o3 nanoparticle, ti2o3, titanium oxide ti2o3, titanium iii oxide nanoparticle, titanium iii oxide trace metals basis 50g
PubChem CID 123111
IUPAC Name oxo(oxotitaniooxy)titanium
SMILES O=[Ti]O[Ti]=O

Specifications

Melting Point 2130°C (decomposition)
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 143.8
Percent Purity 99.8%
Odor Odorless
Density 4.6 g/mL
Assay (metals basis)
Chemical Name or Material Titanium (III) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.