missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(III) fluoride, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagent
Specifications
Specifications
| Boiling Point | 1400°C |
| Melting Point | 1200°C |
| Color | Brown to Red |
| Physical Form | Crystalline Powder |
| Quantity | 5 g |
| Assay Percent Range | 97.5% min. (ex F) |
| Linear Formula | TiF3 |
| Solubility Information | Solubility in water: hydrolyses |
| Formula Weight | 104.9 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
Product Identifier
- Titanium(III) fluoride
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH032-Contact with acids liberates very toxic gas.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction