missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) fluoride, 98%, Thermo Scientific™

Product Code. 10380564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10380564 25 g 25g
10320854 1 g 1g
10445275 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10380564 Supplier Thermo Scientific Acros Supplier No. 384520250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13470-08-1
Molecular Formula F3Ti
Molecular Weight (g/mol) 104.9
MDL Number MFCD00042544

Specifica

Boiling Point 1400°C
Melting Point 1200°C
Color Brown to Red
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 97.5% min. (ex F)
Linear Formula TiF3
Solubility Information Solubility in water: hydrolyses
Formula Weight 104.9
Percent Purity 98%
Identification Confirmed
Packaging Plastic bottle
Chemical Name or Material Titanium(III) fluoride
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Titanium(III) fluoride
Avvertenza
  • Danger
Categoria di pericolo
  • Skin corrosion/irritation Category 1 B
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH032-Contact with acids liberates very toxic gas.
Consigli di prudenza
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.