missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) chloride tetrahydrofuran complex, 97%

Product Code. 10598663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10598663 10 g Glass Bottle 10g
10525983 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10598663 Supplier Thermo Scientific Acros Supplier No. 375160100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18039-90-2 | C12H24Cl3O3Ti | 370.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18039-90-2
Molecular Formula C12H24Cl3O3Ti
Molecular Weight (g/mol) 370.56
MDL Number MFCD01631276
InChI Key MIDYWIPTCYRMQF-UHFFFAOYSA-K
Synonym oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3
PubChem CID 6097232
IUPAC Name oxolane;trichlorotitanium
SMILES C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl

Specifications

Melting Point >250°C
Color Blue to Blue-Green
Physical Form Crystals or Powder
Quantity 10 g
Assay Percent Range 96% min. (Argentometry)
Solubility Information Solubility in water: reacts violently
Formula Weight 370.56
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Titanium(III) chloride tetrahydrofuran complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(III) chloride tetrahydrofuran complex
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.