missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) Chloride, Technical, Solution 12.5% (w/w) TiCl3, Fisher Chemical

Product Code. 10080460 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
10080460 500 mL 500mL
1 options
This item is not returnable. View return policy

Product Code. 10080460

Brand: Fisher Chemical T/1950/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Purple
CAS 7705-07-9
Molecular Formula Cl3Ti
MDL Number MFCD00011266
InChI Key YONPGGFAJWQGJC-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Ti+3]
IUPAC Name titanium(3+) trichloride
Molecular Weight (g/mol) 154.22
Packaging Plastic Coated Glass bottle
pH 1
Quantity 500 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium (III) chloride solution in HCl (12.5% w/w TiCl3)
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Titanium trichloride, Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.