missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%

Product Code. 11314257
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314257 500 g 500g
11304257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314257 Supplier Thermo Scientific Alfa Aesar Supplier No. 036729.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a catalyst in the production of polypropylene. | CAS: 12003-13-3 | AlCl12Ti3 | 595.98 g/mol

Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5% is used as a catalyst in the production of polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-13-3
Molecular Formula AlCl12Ti3
Molecular Weight (g/mol) 595.98
MDL Number MFCD00145412
InChI Key KSKPGAGLAZXNEO-UHFFFAOYSA-H
IUPAC Name titanium(4+) aluminium(3+) heptachloride
SMILES Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN3260
Synonym Titanium aluminum chloride,Aluminum titanium chloride
Solubility Information Insoluble in water.
Formula Weight 596.15
Percent Purity 76.0 to 78.5% (TiCl3)
Packaging Glass bottle
Sensitivity Air and moisture sensitive
Chemical Name or Material Titanium(III) chloride-Aluminum(III) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.