missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
£32.00 - £997.00
Chemical Identifiers
CAS | 12137-20-1 |
---|---|
Molecular Formula | OTi |
Molecular Weight (g/mol) | 63.87 |
MDL Number | MFCD00036279 |
InChI Key | OGIDPMRJRNCKJF-UHFFFAOYSA-N |
Synonym | titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis |
PubChem CID | 61685 |
IUPAC Name | oxotitanium |
SMILES | O=[Ti] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334889
|
Thermo Scientific Alfa Aesar
077126.06 |
5 g |
£32.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324889
|
Thermo Scientific Alfa Aesar
077126.14 |
25 g |
£97.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344889
|
Thermo Scientific Alfa Aesar
077126.22 |
100 g |
£261.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354889
|
Thermo Scientific Alfa Aesar
077126.36 |
500 g |
£997.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12137-20-1 | |
63.87 | |
OGIDPMRJRNCKJF-UHFFFAOYSA-N | |
61685 | |
O=[Ti] |
OTi | |
MFCD00036279 | |
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis | |
oxotitanium |
Specifications
>3000°C | |
12137-20-1 | |
99.5%, metal basis | |
MFCD00036279 | |
Soluble in concentrated sulfuric acid,concentrated hydrofluoric acid and alkali. Insoluble in water,diluted sulfuric acid,hydrochloric acid,nitric acid and organic solvents. | |
O=[Ti] | |
63.87 | |
63.9 | |
Titanium(II) Oxide |
1700°C | |
Powder | |
OTi | |
titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis | |
OGIDPMRJRNCKJF-UHFFFAOYSA-N | |
oxotitanium | |
61685 | |
4.95 |
Safety and Handling
missing translation for 'einecsNumber' : 235-236-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only