missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) hydride, 99% (metals basis)

Product Code. 11317597
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 x 1 kg
Unit Size:
100g
2kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317597 2 x 1 kg 2kg
11327597 5 g 5g
11307597 100 g 100g
11337597 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11317597 Supplier Thermo Scientific Alfa Aesar Supplier No. 012857.D3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Widley used as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. | CAS: 7704-98-5 | H2Ti

Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-98-5
Molecular Formula H2Ti
MDL Number MFCD00151301

Specifications

Melting Point 450°C (decomposition)
Physical Form Powder
Quantity 2 x 1 kg
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in water.
Formula Weight 49.92
Sensitivity Air and moisture sensitive
Density 3.9 g/mL
Chemical Name or Material Titanium(II) hydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.