missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(II) hydride, 99% (metals basis), Thermo Scientific Chemicals
18.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2 x 1 kg
Description
Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 450°C (decomposition) |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in water. |
Formula Weight | 49.92 |
Sensitivity | Air and moisture sensitive |
Density | 3.9 |
Chemical Name or Material | Titanium(II) hydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'