missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Ii) Hydride, 98%, Honeywell Fluka™

Product Code. p-7120876 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15631900 100 g Poly Bottle 100g
15641900 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15631900 Supplier Honeywell-Fluka Supplier No. 209279100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

−325 mesh, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-98-5
MDL Number MFCD00151301

Specifications

Melting Point 400°C
Color Gray
Quantity 100 g
Linear Formula TiH2
UN Number UN1871
Formula Weight 49.88g/mol
Percent Purity 98%
Ignition Point 437°F
Packaging Poly Bottle
Appearance Powder
Density 3.91g/mL at 25°C
Chemical Name or Material Titanium(II) Hydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.