missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)

Product Code. 11338757
Change view
Click to view available options
Quantity:
23 x 25 cm
23 x 100 cm
23 x 500 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338757 23 x 100 cm Each
11328757 23 x 25 cm Each
11348757 23 x 500 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338757 Supplier Thermo Scientific Alfa Aesar Supplier No. 035881.WG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 8.3g/23 x 25cm | CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Melting Point 1660°C
Boiling Point 3287°C
Appearance Silver Gray foil
Density 4.506 g/cm3
Quantity 23 x 100 cm
Odor Odorless
Assay Percent Range 99.7% (Metals basis)
Form Foil
Chemical Name or Material Titanium, Annealed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.