missing translation for 'onlineSavingsMsg'
Learn More

Titanium Dioxide, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15598264 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15598264 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598264 Supplier Avantor J.T.Baker Supplier No. 416201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 13463-67-7
Quantity 500 g
Molecular Formula O2Ti
MDL Number MFCD00011269 MFCD00210650
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES O=[Ti]=O
IUPAC Name dioxotitanium
Molecular Weight (g/mol) 79.87
PubChem CID 26042
ChEBI CHEBI:32234
Formula Weight 79.9
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.26kg
Chemical Name or Material Titanium Dioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.