missing translation for 'onlineSavingsMsg'
Learn More

Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol, Thermo Scientific Chemicals

Catalog Number 11433760
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
17927-72-9
C16H28O6Ti
MFCD00000031
Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)
This item is not returnable. View return policy
17927-72-9
C16H28O6Ti
MFCD00000031
Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)

Titanium(diisopropoxide) bis(2,4-pentanedionate) is used as catalyst, used in sol-gel, coating, primer and crosslinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Physical Form Liquid
Quantity 100 g
UN Number UN1219
Flash Point 18°C (61°F)
Solubility Information Hydrolyze with water.
Formula Weight 364.3
Concentration or Composition (by Analyte or Components) 75% in isopropanol
Odor Alcohol-like
Refractive Index 1.494
Sensitivity Moisture sensitive
Density 1.010 g/mL
Chemical Name or Material Titanium (diisopropoxide) bis (2,4-pentanedionate)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'