missing translation for 'onlineSavingsMsg'
Learn More

Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol

Product Code. 11423760
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423760 500 g 500g
11433760 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423760 Supplier Thermo Scientific Alfa Aesar Supplier No. 089360.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17927-72-9 | C16H28O6Ti

Titanium(diisopropoxide) bis(2,4-pentanedionate) is used as catalyst, used in sol-gel, coating, primer and crosslinking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17927-72-9
Molecular Formula C16H28O6Ti
MDL Number MFCD00000031
Synonym Titanium diisopropoxide bis(acetylacetonate); Bis(acetylacetonato)diisopropoxytitanium(IV)

Specifications

Color Brown
Physical Form Liquid
Quantity 500 g
UN Number UN1219
Flash Point 18°C (61°F)
Solubility Information Hydrolyze with water.
Formula Weight 364.3
Concentration or Composition (by Analyte or Components) 75% in isopropanol
Odor Alcohol-like
Refractive Index 1.494
Sensitivity Moisture sensitive
Density 1.010 g/mL
Chemical Name or Material Titanium (diisopropoxide) bis (2,4-pentanedionate)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(diisopropoxide) bis(2,4-pentanedionate), 75% in isopropanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Titanium(diisopropoxide) bis(2,4-pentanedionate), isopropanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.