missing translation for 'onlineSavingsMsg'
Learn More

Titanium carbide, nanopowder

Product Code. 15442077
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442077 25 g 25g
15432077 5 g 5g
15452077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442077 Supplier Thermo Scientific Alfa Aesar Supplier No. 044922.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-08-5 | CTi | 59.88 g/mol

Titanium carbide is used in the manufacture of wear-resistant tools, cutting tools, As a coating for abrasive steel bearings, In the form of nano titanium carbide ceramic in optics applications, Enhance the conductivity of materials and as a nucleating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
Molecular Weight (g/mol) 59.88
MDL Number MFCD00011268
InChI Key YXIVWSJCLXKLJL-UHFFFAOYSA-N
IUPAC Name methanidylidynetitaniumylium
SMILES [C-]#[Ti+]

Specifications

Boiling Point 4820°C
Melting Point ∽3140°C
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in nitric acid and aqua regia. Insoluble in water.
Formula Weight 59.91
Density 4.93 g/mL
Chemical Name or Material Titanium carbide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.