missing translation for 'onlineSavingsMsg'
Learn More

Titanium carbide, 99.5% (metals basis)

Product Code. 11349307
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349307 1 kg 1kg
11359307 50 g 50g
11339307 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349307 Supplier Thermo Scientific Alfa Aesar Supplier No. 040178.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-08-5 | CTi | 59.88 g/mol

Used in preparation of cermets and abrasion-resistant surface coating on metal parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
Molecular Weight (g/mol) 59.88
MDL Number MFCD00011268
InChI Key YXIVWSJCLXKLJL-UHFFFAOYSA-N
IUPAC Name methanidylidynetitaniumylium
SMILES [C-]#[Ti+]

Specifications

Boiling Point 4820°C
Melting Point ∽3140°C
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 59.91
Density 4.93
Chemical Name or Material Titanium carbide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.