Learn More
Diversey™ Titan Chlor Tablets
Product Code. 17663095
Change view
Click to view available options
Quantity:
6/Cs.
Unit Size:
Pack of 600
Product Code. Quantity Unit Size
17663095 6/Cs. Pack of 600
1 options
Product Code. 17663095 Supplier Diversey™ Supplier No. 101100211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Titan Chlor-Tabs 2500 are a blend of NaDCC, effervescent agents, and tableting agents.
They provide an accurate and convenient means of making up solutions of known available chlorine strengths for a variety of disinfection tasks. Titan Chlor- Tabs 2500 are soluble in warm or cold water and are compatible when dissolved with neutral detergents for making a one stage sanitizing solution.
  • Contains 4.5 g Troclosene
  • Chlorine release agents have been shown to be effective against a wide range of micro-organisms: bacteria, yeast, fungi, viruses, spores. Efficacy confirmed against European norms.
  • For increased safety in general cleaning of high risk areas (theatres, maternity units, etc.) drains and sinks.
Specifications
Sterility Non-sterile
Composition Chlorine based tablets (4.5 g Troclosene)
Description For GB only
Quantity 6/Cs.