missing translation for 'onlineSavingsMsg'
Learn More

Tiron, Indicator, Honeywell Fluka™

Product Code. p-7120875 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 15627390

Brand: Honeywell-Fluka 8946010G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00149531
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Specifications

Melting Point 300°C
Linear Formula (OH)2C6H2(SO3Na)2 · H2O
UN Number NONH for all modes of transport
Solubility Information 1350g/L
Formula Weight 332.22g/mol
Packaging Plastic bottle
Color Violet to White
pH 5.0
Quantity 10 g
Physical Form Solid
Chemical Name or Material Tiron
Show More Show Less
compliance-icons
Product Identifier
  • Tiron
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4,5-Dihydroxy-1,3-benzenedisulfonic acidhydrate disodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.