missing translation for 'onlineSavingsMsg'
Learn More

Tiratricol, MedChemExpress

Product Code. 16468676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468676 Supplier Medchem Express Supplier No. HYB1201
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tiratricol is a thyroid hormone analog with hepatic, has been used to suppress pituitary TSH secretion, with attenuation of extrapituitary thyromimetic effects.

Tiratricol is a thyroid hormone analog with hepatic, has been used to suppress pituitary TSH secretion, with attenuation of extrapituitary thyromimetic effects.

Specifications

Chemical Name or Material Tiratricol
CAS 51-24-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H9I3O4
Quantity 100 mg
Synonym 3,3',5-Triiodothyroacetic acid
Solubility Information DMSO : ≥ 42 mg/mL (67.53 mM)
SMILES O=C(O)CC1=CC(I)=C(OC2=CC=C(O)C(I)=C2)C(I)=C1
Molecular Weight (g/mol) 621.93
Formula Weight 621.93
Percent Purity 98.02%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.