missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tirapazamine, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Description
Tirapazamine is an anticancer agent that shows selective cytotoxicity for hypoxic cells in solid tumors, thereby inducing single-and double-strand breaks in DNA, base damage, and cell death. Tirapazamine is an anticancer and bioreductive agent.
Specifications
Specifications
| Chemical Name or Material | Tirapazamine |
| Physical Form | Solid |
| CAS | 27314-97-2 |
| Color | Orange Red |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C7H6N4O2 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | SR259075 SR4233 Win59075 SML 0552 SR 259075 Tirazone |
| Solubility Information | DMSO : 62.5 mg/mL (350.83 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction