missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIRAP Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818052
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818052 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818052 Supplier Invitrogen™ Supplier No. PA520028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human heart tissue lysate. PA5-20028 can be used with blocking peptide PEP-0147.

Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. These proteins act through adaptor molecules such as TIRAP and MyD88 to activate various kinases and transcription factors. In TIRAP-deficient mice, TLR signaling in response to TLR2 ligands (using either TLR1 and TLR6 as co-receptors) is totally abolished, suggesting that MyD88 and TIRAP work together and are both required for TLR2 signaling. Furthermore, these mice are also resistant to the toxic effects of LPS and show defects in NF-βB and MAP kinase activation, suggesting that TIRAP is also involed in TLR4 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIRAP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TIRAP
Gene Accession No. P58753, Q99JY1
Gene Alias AA407980; adapter protein wyatt; adaptor protein Wyatt; BACTS1; C130027E04Rik; FLJ42305; HGNC:17192; Mal; myD88 adapter-like protein; MyD88-2; OTTHUMP00000179130; TIR domain containing adaptor protein; TIR domain-containing adapter protein; TIRAP; Tlr4ap; toll/interleukin-1 receptor domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; toll-interleukin 1 receptor (TIR) domain-containing adaptor protein; Toll-like receptor 4 adaptor protein; Toll-like receptor adaptor protein; Wyatt
Gene Symbols TIRAP
Host Species Rabbit
Immunogen A peptide corresponding to 15 amino acids near the C-terminus of mouse TIRAP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114609, 117149, 680127
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.