missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPRL Antibody (OTI3D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74529CL1
This item is not returnable.
View return policy
Description
TIPRL Monoclonal antibody specifically detects TIPRL in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| TIPRL | |
| Monoclonal | |
| CoraFluor 1 | |
| dJ69E11.3, MGC3794, putative MAPK activating protein, Putative MAPK-activating protein PM10, TIP41, TOR signaling pathway regulator-like (S. cerevisiae), TIP41-like protein, TIP41Type 2A-interacting protein, TIPTIP41, TOR signalling pathway regulator-like (S. cerevisiae) | |
| Full length human recombinant protein of human TIPRL (NP_690866) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI3D11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 261726 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction