missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPRL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98503-50ul
This item is not returnable.
View return policy
Description
TIPRL Polyclonal antibody specifically detects TIPRL in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TIPRL | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| dJ69E11.3, MGC3794, putative MAPK activating protein, Putative MAPK-activating protein PM10, TIP41, TOR signaling pathway regulator-like (S. cerevisiae), TIP41-like protein, TIP41Type 2A-interacting protein, TIPTIP41, TOR signalling pathway regulator-like (S. cerevisiae) | |
| Produced in rabbits immunized with E. coli-derived Human TIPRL fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 261726 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction