missing translation for 'onlineSavingsMsg'
Learn More

Tipranavir, MedChemExpress

Product Code. 16456328
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16456328

Brand: Medchem Express HY15148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tipranavir (PNU-140690) inhibits the enzymatic activity and dimerization of HIV-1 protease, exerts potent activity against multi-protease inhibitor (PI)-resistant HIV-1 isolates with IC50s of 66-410 nM. Tipranavir inhibits SARS-CoV-2 3CLpro activity.

Tipranavir (PNU-140690) inhibits the enzymatic activity and dimerization of HIV-1 protease, exerts potent activity against multi-protease inhibitor (PI)-resistant HIV-1 isolates with IC50s of 66-410 nM. Tipranavir inhibits SARS-CoV-2 3CLpro activity.

Specifications

Chemical Name or Material Tipranavir
CAS 174484-41-4
Color White
Purity Grade Notes Research
Molecular Formula C31H33F3N2O5S
Quantity 10 mg
Synonym PNU-140690
Solubility Information DMSO : 200 mg/mL (331.86 mM; Need ultrasonic) ∣Ethanol : ≥ 50 mg/mL (82.97 mM)
SMILES O=C1C([C@H](CC)C2=CC=CC(NS(C3=CC=C(C(F)(F)F)C=N3)(=O)=O)=C2)=C(O)C[C@](CCC4=CC=CC=C4)(CCC)O1
Molecular Weight (g/mol) 602.66
Formula Weight 602.66
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.