missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPIN Antibody (OTI2E6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74528CL1
This item is not returnable.
View return policy
Description
TIPIN Monoclonal antibody specifically detects TIPIN in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| TIPIN | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein | |
| Full length human recombinant protein of human TIPIN (NP_060328) produced in E.coli. | |
| 0.1 mL | |
| DNA Repair | |
| 54962 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI2E6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction