missing translation for 'onlineSavingsMsg'
Learn More

TIPIN Antibody, DyLight 550, Novus Biologicals™

Product Code. 18342205 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342205 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342205 Supplier Novus Biologicals Supplier No. NBP298947R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIPIN Polyclonal specifically detects TIPIN in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIPIN
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TIPIN.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 54962
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.