missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tipelukast, MedChemExpress
Description
Tipelukast (KCA 757) is a sulfidopeptide leukotriene receptor antagonist, an orally bioavailable anti-inflammatory agent and used for the treatment of asthma.
Specifications
Specifications
| Chemical Name or Material | Tipelukast |
| CAS | 125961-82-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H38O7S |
| Quantity | 1 mg |
| Synonym | KCA 757 MN 001 |
| Solubility Information | Ethanol : 1 mg/mL (1.88 mM; Need ultrasonic and warming) |
| SMILES | CC(C1=C(O)C(CCC)=C(SCCCOC2=C(CCC)C(OCCCC(O)=O)=C(C(C)=O)C=C2)C=C1)=O |
| Molecular Weight (g/mol) | 530.67 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?