Learn More
TIPE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15940-1-AP-20UL
 View more versions of this product
Product Code. 16875344
  • £135.00 / 20µL
Description
Acts as a negative regulator of innate and adaptive immunity by maintaining immune homeostasis. Negative regulator of Toll-like receptor and T-cell receptor function. Prevents hyperresponsiveness of the immune system and maintains immune homeostasis. Inhibits JUN/AP1 and NF-kappa-B activation. Promotes Fas-induced apoptosis (By similarity).
Specifications
Specifications
TIPE2
Polyclonal
Unconjugated
TNFAIP8L2
Inflammation factor protein 20, TIPE2, TNFAIP8 like protein 2, TNFAIP8L2
Rabbit
Antigen Affinity Chromatography
RUO
310663, 69769, 79626
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AYJ8, Q6P589, Q9D8Y7
Tnfaip8l2
TIPE2 Fusion Protein Ag7964
20 μL
Primary
Mouse, Rat, Human