missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15940-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a negative regulator of innate and adaptive immunity by maintaining immune homeostasis. Negative regulator of Toll-like receptor and T-cell receptor function. Prevents hyperresponsiveness of the immune system and maintains immune homeostasis. Inhibits JUN/AP1 and NF-kappa-B activation. Promotes Fas-induced apoptosis (By similarity).Specifications
| TIPE2 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP8L2 | |
| Inflammation factor protein 20, TIPE2, TNFAIP8 like protein 2, TNFAIP8L2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 310663, 69769, 79626 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AYJ8, Q6P589, Q9D8Y7 | |
| Tnfaip8l2 | |
| TIPE2 Fusion Protein Ag7964 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction